

5-Aza-2'-deoxycytidine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2353-33-5
:
Formula:C₈H₁₂N₄O₄
MDL Number:MFCD00043011
Melting Pt:200 °C
ADR:6.1,III
:228.21 g/mol
:GHS07
:GHS08
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together