(±)-2-Methyltetrahydrofuran, extra dry 99+% stabiliser free, AcroSeal®
:96-47-9
:
:C₅H₁₀O
:MFCD00005367
:79
:–136 °C
:0.86 g/cm³
:86.13 g/mol
:–11 °C (closed cup)
:2536
:3 II
:GHS02
:GHS05
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
- Residue after evaporation:≤0.001 %
- Peroxides:≤20 ppm
Frequently Bought Together