

Ethyl trifluoropyruvate 98+%
Catalog # ACRO399010050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3390 to 1.3410 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
About this item
:
:170.09 g/mol
Flash Pt:27 °C
Storage Temperature:Ambient
UN:3272
ADR:3,III
:GHS07
:GHS02
CAS Number:13081-18-0
Formula:C₅H₅F₃O₃
MDL Number:MFCD00114935