Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
2-Iodobenzonitrile 99%
 
undefined
 
2-Iodobenzonitrile 99%

 

  • Appearance:
    Yellow Solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    50 to 54 °C
  • GC:
    ≥98.5 %

 

 

Frequently Bought Together