Order Entry
Northern Ireland
ContactUsLinkComponent
2-Chloro-5-formylpyridine 97%
Irritant
2-Chloro-5-formylpyridine 97%
Certificates
undefined
Irritant
2-Chloro-5-formylpyridine 97%

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    77 to 82 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together