
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin 95%
ACRO292151000
:
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
- Appearance:Violet to purple Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
:
:974.56 g/mol
:GHS07
:Ambient
:25440-14-6
:C₄₄H₁₀F₂₀N₄
:MFCD00010032