
(1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one 95%, 98% ee
Catalog # ACRO362200010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals or needles
- Infrared spectrum:Conforms
- GC:≥94 %
- Enantiomeric excess:≥97.5 % (GC)
Specification Test Results
About this item
CAS Number:178032-63-8
:
Formula:C₉H₁₃NO₃
MDL Number:MFCD01320865
:183.207 g/mol
Melting Pt:150…152 °C
:GHS07
Storage Temperature:Ambient