

1,1-Di(tert-butylperoxy)cyclohexane 50% in mineral oil
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3006-86-8
:
Formula:C₁₄H₂₈O₄
MDL Number:MFCD00084800
:260.374 g/mol
Boiling Pt:52…54 °C (,1 mmHg)
Density:0,95 g/cm³
Storage Temperature:Refrigerator
UN:3105
ADR:5.2,
:GHS02
:GHS08
Specification Test Results
- Appearance:Clear liquid
- Titration Iodometric:49 to 51 %
- Color scale:≤20 APHA
- Impurity:≤0.2 % Hydroperoxides (as TBHP)
Specification Test Results
Frequently Bought Together