Order Entry
Northern Ireland
ContactUsLinkComponent
(Chloromethoxy)ethane 80% tech. stabilised
Flammable
Irritant
Health
(Chloromethoxy)ethane 80% tech. stabilised
Catalog # ACRO355720250
CAS Number:  
undefined
Flammable
Irritant
Health
(Chloromethoxy)ethane 80% tech. stabilised
Catalog # ACRO355720250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4000 to 1.4050 (20°C, 589 nm)
  • GC:
    ≥75 %
  • Stabilizer:
    CaCl2

Specification Test Results

About this item

 :
 :94.54 g/mol
Boiling Pt:82 °C
Melting Pt:–98 °C
Density:1,01 g/cm³
Flash Pt:19 °C
UN:2354
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
CAS Number:3188-13-4
Formula:ClCH₂OCH₂CH₃
MDL Number:MFCD00000887