

Nicotine ditartrate dihydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6019-06-3
:
Formula:C₁₀H₁₄N₂·2C₄H₆O₆·2H₂O
MDL Number:MFCD00069368
:462.41 g/mol
Melting Pt:97…100 °C
UN:1659
ADR:6.1,II
:GHS06
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Water:5 to 10 % (K.F.)
- Titration with TBAH:≥97.5 % (On dry substance)
Specification Test Results
Frequently Bought Together