Order Entry
Northern Ireland
ContactUsLinkComponent
6,7-Dihydrobenzo[b]thiophen-4-one 97%
Irritant
6,7-Dihydrobenzo[b]thiophen-4-one 97%
Catalog # ACRO122890010
CAS Number:  
undefined
Irritant
6,7-Dihydrobenzo[b]thiophen-4-one 97%
Catalog # ACRO122890010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to yellow Low melting solid or liquid after melting
  • Infrared spectrum:
    Conforms
  • Melting point:
    34 to 40 °C
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :152.211 g/mol
Boiling Pt:120 °C (10 mmHg)
Flash Pt:110 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:13414-95-4
Formula:C₈H₈OS
MDL Number:MFCD00005861