Order Entry
Northern Ireland
ContactUsLinkComponent
 
(tert-Butoxycarbonylmethylene)triphenylphosphorane 97%
 
undefined
(tert-Butoxycarbonylmethylene)triphenylphosphorane 97%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    146 to 155 °C
  • Carbon (C):
    73.5 to 79.5 %

 

 

Frequently Bought Together