N-Vinylphthalimide 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3485-84-5
:
Formula:C₁₀H₇NO₂
MDL Number:MFCD00078446
:173.17 g/mol
Melting Pt:84…88 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:84 to 88 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together