

(1-Ethoxycyclopropoxy)trimethylsilane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27374-25-0
:
Formula:C₈H₁₈O₂Si
MDL Number:MFCD00074986
:174.315 g/mol
Boiling Pt:50…53 °C (22 mmHg)
Density:0,867 g/cm³
Flash Pt:26 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4060 to 1.4090 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together