Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid 97%
 
undefined
 
6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid 97%
 

 

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    187 to 194 °C
  • Titration with NaOH:
    ≥96.0 %

 

 

Frequently Bought Together