1,4-Bis(2-hydroxyethoxy)benzene 98+%
:104-38-1
:
:C₁₀H₁₄O₄
:MFCD00002861
:198.22 g/mol
:185…200 °C (,3 mmHg)
:104…107 °C
:Ambient
- Appearance:White to almost white Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:100 to 107 °C
- GC:≥98.0 %
Frequently Bought Together