ContactUsLinkComponent
Phenyl phosphate, disodium salt dihydrate 98%
Phenyl phosphate, disodium salt dihydrate 98%
Certificates
undefined
Phenyl phosphate, disodium salt dihydrate 98%
Certificates
Specification Test Results
  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    12 to 16 % (K.F.)
  • Titration with HClO4:
    ≥97.5 %