



Chrysoidine, pure
Catalog # ACRO200740500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Appearance:Bordeaux to purple Crystalline powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Visual transition interval:from pH 4.0 (orange) to pH 7.0 (yellow)
Specification Test Results
About this item
CAS Number:532-82-1
:
Formula:C₁₂H₁₃ClN₄
MDL Number:MFCD00012976
:248.714 g/mol
Melting Pt:235 °C
Storage Temperature:Ambient
UN:3077
ADR:9,III
:GHS08
:GHS05
:GHS09
:GHS07