

1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene 90%
Catalog # ACRO199550050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4530 to 1.4570 (20°C, 589 nm)
- GC:≥88 %
Specification Test Results
About this item
CAS Number:59414-23-2
:
Formula:C₈H₁₆O₂Si
MDL Number:MFCD00008498
Boiling Pt:68…69 °C (14 mmHg)
Density:0,88 g/cm³
Flash Pt:45 °C
UN:1993
ADR:3,III
:172.30 g/mol
:GHS02
:GHS07
Storage Temperature:Refrigerator