Order Entry
Northern Ireland
ContactUsLinkComponent
 
Polyethylene glycol, M.W. 250,000 - 400,000
 
undefined
Polyethylene glycol, M.W. 250,000 - 400,000

 

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 % (10 g, 50°C, 5 h, 10 mmHg)
  • Viscosity:
    2000 to 4000 mPa.s (10 % w/w in water at 25°C)

 

 

Frequently Bought Together