1-(3-aminopropyl)pyrrolidin-2-one 95%
Catalog # ACRO159600050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4970 to 1.4990 (20°C, 589 nm)
- GC:≤3 % 2-Pyrrolidinone
Specification Test Results
About this item
:
:142.202 g/mol
Boiling Pt:120…123 °C (1 mmHg)
Density:1,01 g/cm³
Flash Pt:>112 °C
Storage Temperature:Ambient
UN:3267
ADR:8,II
:GHS05
CAS Number:7663-77-6
Formula:C₇H₁₄N₂O
MDL Number:MFCD00003201