Order Entry
Northern Ireland
ContactUsLinkComponent
Cresol red sodium salt, water soluble, pure indicator
Cresol red sodium salt, water soluble, pure indicator
Certificates
undefined
Cresol red sodium salt, water soluble, pure indicator

Specification Test Results

  • Appearance:
    Brown Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.1 % in water) Clear
  • Visual transition interval:
    from pH 7.0 (yellow) to pH 8.8 (purple)

Specification Test Results

Related Information

Frequently Bought Together