
5-Norbornene-2-carboxaldehyde (endo + exo mixture) 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5453-80-5
:
Formula:C₈H₁₀O
MDL Number:MFCD00167568
:122.17 g/mol
Boiling Pt:67…70 °C (12 mmHg)
Density:1,018 g/cm³
Flash Pt:50,6 °C
Storage Temperature:Ambient
UN:1989
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4870 to 1.4920 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together