Order Entry
Northern Ireland
ContactUsLinkComponent
cis-1,2,3,6-Tetrahydrophthalimide 96%
Irritant
cis-1,2,3,6-Tetrahydrophthalimide 96%
Certificates
undefined
Irritant
cis-1,2,3,6-Tetrahydrophthalimide 96%

Specification Test Results

  • Appearance:
    Yellow to light beige to light brown Powder or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    131 to 138 °C
  • Titration with NaOH:
    ≥95 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information

Frequently Bought Together