
2-Acetyl-6-methoxynaphthalene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3900-45-6
:
Formula:C₁₃H₁₂O₂
MDL Number:MFCD00021643
:200.24 g/mol
Melting Pt:108…109 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to beige Powder
- Infrared spectrum:Conforms
- Melting point:106 to 109 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together