Order Entry
Northern Ireland
ContactUsLinkComponent
trans-Stilbene oxide 99%
trans-Stilbene oxide 99%
Certificates
undefined
trans-Stilbene oxide 99%

Specification Test Results

  • Appearance:
    White Crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    67 to 70 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together