
Humic acid sodium salt 45 - 70% (as humic acid), crystalline powder, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:68131-04-4
:
Formula:C₉H₈Na₂O₄
MDL Number:MFCD00135560
:226.139 g/mol
Melting Pt:>300 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Black Crystalline powder
- Infrared spectrum:Conforms
- Assay:45 to 70 % (as Humic acid)
- Loss on drying:14 % to 25 % (105°C, 4 h)
Specification Test Results
Frequently Bought Together