
L-Histidine methyl ester dihydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7389-87-9
:
Formula:C₇H₁₁N₃O₂
MDL Number:MFCD00012701
:242.10 g/mol
Melting Pt:207 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 %
- HPLC:≥96.0 %
- Specific optical rotation:+8° to +11° (20°C, 589 nm) (c=2, water)
Specification Test Results
Frequently Bought Together