
1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimidemetho-p-toluenesulphonate 95%
:2491-17-0
:
:C₂₁H₃₃N₃O₄S
:MFCD00011979
:423.57 g/mol
:110…116 °C
:GHS07
:Ambient
- Appearance:White to light yellow Powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:112 to 120 °C
- HPLC:≥94 %
Frequently Bought Together