
N-(4-Vinylbenzyl)-N,N-dimethylamine 90% stabilised, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2245-52-5
:
Formula:C₁₁H₁₅N
MDL Number:MFCD00078270
:161.25 g/mol
Boiling Pt:72…74 °C (1 mmHg)
Density:0,92 g/cm³
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear yellow to orange to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5360 to 1.5400 (20°C, 589 nm)
- GC:≥88 %
- Stabilizer:0.1 % Hydroquinone
Specification Test Results
Frequently Bought Together