Sodium methoxide 0.5 M in methanol, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sodium methylate, Sodium methanolate
CAS Number:124-41-4
:
Formula:CH₃NaO
MDL Number:MFCD00012179
Density:0.8 g/cm³
Molecular Weight:54.02 g/mol
Melting Pt:–98 °C
Flash Pt:11 °C
UN:1992
ADR:3 II
:GHS06
:GHS08
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Strength:0.45 to 0.60 M
- GC:≥95.0 % (sum of cis and trans)
Specification Test Results
Frequently Bought Together