Paraffin wax, granular, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:8002-74-2
:8042-47-5
Boiling Pt:322 °C (760 mmHg)
Melting Pt:58…62 °C
Flash Pt:198 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Pellets
- Infrared spectrum:Conforms
- Congealing point:58 °C to 62 °C
- Titration Argentometric:≥98.5 %
Specification Test Results
Frequently Bought Together