
4-(4-Ethoxyphenylazo)-m-phenylenediamine, mixture of mono and dihydrochloride mixture of mono- and di-hydrochloride 98%
ACRO409680100
:
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Appearance:Dark brown to black Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- GC:≥97.5 %
:
:215 °C
:GHS07
:Ambient
:74367-87-6
:C₁₄H₁₆N₄O·xHCl
:MFCD00042023