ContactUsLinkComponent
4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran 96+%
Flammable
Irritant
4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran 96+%
Certificates
undefined
Flammable
Irritant
4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran 96+%
Specification Test Results
  • Appearance:
    Red to red-brown Powder or crystals
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %
  • UV Lambda max:
    479 to 483 nm (in DMSO)
  • Molar absorption coefficient:
    ≥42000 l/(mol.cm) (at Lambda max)