
6,9-Diamino-2-ethoxyacridine lactate monohydrate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6402-23-9
:
Formula:C₁₈H₂₃N₃O₅
MDL Number:MFCD00149646
:361.39 g/mol
Melting Pt:243…245 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥94 %
Specification Test Results
Frequently Bought Together