Cellulose acetate butyrate 50 - 54% (butyryl content)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9004-36-8
:
Formula:C₈₄H₁₃₀O₄₉
MDL Number:MFCD00081513
:1923.90 g/mol
Melting Pt:127…240 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline or granular powder
- Infrared spectrum:Conforms
- Viscosity:0.01 to 0.03 s
- Average molecular weight:16000 (Mn)
- Acetyl content:≤4 %
- Hydroxyl content:1.5 % (typical)
- Butyryl content:50 to 54 %
Specification Test Results
Frequently Bought Together