
Bis(2-hydroxyethyl)dimethylammonium chloride 99%
ACRO402530250
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Colorless to white to off-white Adhering crystalline powder or crystals
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 %
- GC:≥97.5 %
:
:169.65 g/mol
:GHS07
:Ambient
:38402-02-7
:C₆H₁₆ClNO₂
:MFCD00041987