DL-Alanine ethyl ester hydrochloride 99%
Catalog # ACRO400430250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White Powder and/or granules
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 %
- GC:≥97.5 %
Specification Test Results
About this item
:
Melting Pt:87 °C
:153.606 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:617-27-6
Formula:C₅H₁₂ClNO₂
MDL Number:MFCD00013018