Order Entry
Northern Ireland
ContactUsLinkComponent
D-ɑ-Tocopherylquinone 90-95%
D-ɑ-Tocopherylquinone 90-95%
Catalog # ACRO350305000
CAS Number:  
undefined
D-ɑ-Tocopherylquinone 90-95%
Catalog # ACRO350305000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Clear amber to red Viscous liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥90 %

Specification Test Results

About this item

 :
 :446.72 g/mol
Flash Pt:>110 °C
Storage Temperature:Refrigerator
CAS Number:7559-04-8
Formula:C₂₉H₅₀O₃
MDL Number:MFCD00135657