Sodium sulphanilate hydrate 97%
Catalog # ACRO346882500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Appearance:White to light brown to pink Crystalline powder
- Infrared spectrum:Conforms
- Assay:≥96.0 % (on dry substance)
- Water:≤16 % (K.F.)
Specification Test Results
About this item
:
:195.17 g/mol
Melting Pt:288 °C
Storage Temperature:Ambient
CAS Number:123333-70-0
Formula:C₆H₇NO₃S
MDL Number:MFCD00064394