
(+)-Diacetyl-L-tartaric anhydride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6283-74-5
:
Formula:C₈H₈O₇
MDL Number:MFCD00037918
:216.15 g/mol
Melting Pt:130…135 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White Powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:130 to 135 °C
- Specific optical rotation:+56° to +62° (20 °C, 589 nm) (c=6, acetone)
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together