
Bafilamycin A1 95%
Catalog # ACRO328120001
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:0,1 mg
- Seq:1286
Specifications
Specification Test Results
- Appearance:White to off-white Powder or solid
- HPLC:≥94 %
Specification Test Results
About this item
:
:622.83 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:88899-55-2
Formula:C₃₅H₅₈O₉
MDL Number:MFCD06795130