
DAB EASY 10 mg, tablets for immunohistochemistry
Catalog # ACRO328005000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:Light grayish white Tablet
- Assay:9 to 11 mg/tablet
- pH:6.0 to 7.5
Specification Test Results
About this item
:
:360.11 g/mol
:GHS08
Storage Temperature:Freezer
CAS Number:7411-49-6
Formula:C₁₂H₁₈Cl₄N₄
MDL Number:MFCD00007725