
4-Methoxybenzyl isocyanate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56651-60-6
:
Formula:C₉H₉NO₂
MDL Number:MFCD00673064
:163.176 g/mol
Boiling Pt:90 °C (,6 mmHg)
Storage Temperature:Ambient
UN:2206
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5305 to 1.5325 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together