

Niobium(V) ethanolate 99.95% (trace metals basis)
ACRO316880050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Yellow to orange-brown Liquid
- Total trace metal impurities:≤500 ppm
:
:140…142 °C (,1 mmHg)
:6 °C
:1,268 g/cm³
:36 °C
:Ambient
:2920
:8,II
:318.209 g/mol
:GHS05
:GHS02
:3236-82-6
:C₁₀H₂₅NbO₅
:MFCD00015122