3,5-Bis(trifluoromethyl)phenyl Isocyanate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Isocyanato-3,5-bis(trifluoromethyl)benzene, 3,5-Bis(trifluoromethyl)phenylisocyanate, Isocyanic Acid 3,5-Bis(trifluoromethyl)phenyl Ester
CAS Number:16588-74-2
:
Formula:C₉H₃F₆NO
MDL Number:MFCD00013559
Boiling Pt:62 °C (2 mmHg)
Density:1.47 g/cm³
Molecular Weight:255.12 g/mol
Flash Pt:67 °C
Storage Temperature:Ambient
UN:2206
ADR:6.1 III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4295 to 1.4315 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together