

Sodium metatungstate in water stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12141-67-2
:
Formula:H₂Na₆O₄₀W₁₂H₂
MDL Number:MFCD00150663
:2986.13 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear yellow to light green Liquid
- Assay:≥67 % (WO3)
- Stabilizer:Typically 0.02 % Na2Cr2O7 salt
Specification Test Results
Frequently Bought Together