Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
L-Proline methyl ester hydrochloride 98%
 
undefined
 
L-Proline methyl ester hydrochloride 98%

 

  • Appearance:
    White Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥97.5 %
  • Water:
    ≤0.5 % (K.F.)
  • Specific optical rotation:
    -31° to -35° (20°C, 589 nm) (c=1, H2O)

 

 

Frequently Bought Together