
cis-2-Amino-1-cyclopentanecarboxylic acidhydrochloride hemihydrate 99%
Catalog # ACRO298382500
Supplier: Thermo Fisher Scientific
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Water:≤6 % (K.F.)
- Titration with HClO4:≥98.5 % (On dry substance)
Specification Test Results
About this item
:
:183.632 g/mol
Melting Pt:199…202 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:212755-84-5
Formula:C₆H₁₄ClNO₃
MDL Number:MFCD00800502