Order Entry
Northern Ireland
ContactUsLinkComponent
Potassium tris(1-pyrazolyl)borohydride 93%
Irritant
Flammable
Potassium tris(1-pyrazolyl)borohydride 93%
Catalog # ACRO296010050
CAS Number:  
undefined
Irritant
Flammable
Potassium tris(1-pyrazolyl)borohydride 93%
Catalog # ACRO296010050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    182 to 188 °C
  • Titration with HClO4:
    ≥91 %

Specification Test Results

About this item

 :
 :252.13 g/mol
Melting Pt:182…188 °C
Storage Temperature:Ambient
UN:2813
ADR:4.3,III
 :GHS07
 :GHS02
CAS Number:18583-60-3
 :MFCD03414149
Formula:C₉H₁₀BKN₆
MDL Number:MFCD00040346